missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM-Eagle without L-Glutamine

Product Code. 31908490 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908490 100 mL 100mL
31908415 500 mL 500mL
31908407 10 x 500 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31908490 Supplier Sartorius Supplier No. 010251B
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM is a modification of BME with higher concentrations of essential nutrients for cultivating cells grown in monolayers.

  • MEM has been utilized for the cultivation of a wide array of cells grown in monolayers.
  • The optional supplementation of non-essential Amino Acids (NEAA) to the formulations that incorporate either Hank's or Earle's Salts has broadened the usefulness of this medium.
  • We offer a variety of MEM modifications for a range of cell culture applications.

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Earle's Balanced Salt Solution (EBSS), Phenol Red, Sodium Bicarbonate
Without Additives No Glutamine
Quantity 100 mL
Culture Type Monolayer Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.