missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM Eagle Non-Essential Amino Acids Solution

Product Code. 31908354 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
10mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908354 100 mL 10mL
31908350 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31908354 Supplier Sartorius Supplier No. 013401B
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

MEM Non-Essential Amino Acids Solution (100X) containing 10 mM concentration of each non-essential amino acid.

  • Modified as follows:
    • With L-ALANINE
    • With L-ASPARAGINE.H2O
    • With L-ASPARTIC ACID
    • With L-GLUTAMIC ACID
    • With GLYCINE
    • With L-PROLINE
    • With L-SERINE

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Proline
Without Additives None
Quantity 100 mL
Culture Type Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.