missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM-Eagle Amino Acids Solution 50X

Product Code. 31908454 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908454 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908454 Supplier Sartorius Supplier No. 013251B
 Restricted Item
Estimated Shipment: 25-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for cell culture, allowing the growth and propagation of a wide spectrum of cell types under controlled conditions.

  • The solution undergoes extensive and vigorous Quality Control Protocols.

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 100 mL
Culture Type Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.