missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM-Alpha

Product Code. 31908494 Shop All Sartorius Products
Change view
Click to view available options
With Additives:
High Glucose, Glutamine
Low Glucose, Glutamine, Sodium Bicarbonate, Phenol Red
Unit Size:
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. With Additives Unit Size
31908494 High Glucose, Glutamine 500mL
31908487 Low Glucose, Glutamine, Sodium Bicarbonate, Phenol Red 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31908494 Supplier Sartorius Supplier No. 010431A
 Restricted Item
Estimated Shipment: 17-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Minimum Essential Medium-α is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells.

  • MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells.

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives High Glucose, Glutamine
Without Additives No Ribonucleosides
Culture Type Monolayer Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.