Learn More
BUCHI Melting and Boiling Point Instruments
Product Code. p-9455007 Shop All Buchi Products
Change view
Click to view available options
Product Type:
Automatic Melting Point Instrument
Manual Melting Point Apparatus
Includes:
Melting Point instrument
Melting Point instrument, Sample Loader M-569, Calibration Set M-565, melting point tubes (100), boiling point tubes (10), boiling capillary (10), sample holder, cleaning tool, CD demo melting point monitor, protective cover, USB cable (2m)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Includes unitSize
10499094 Manual Melting Point Apparatus Melting Point instrument Each
17917990 Automatic Melting Point Instrument Melting Point instrument, Sample Loader M-569, Calibration Set M-565, melting point tubes (100), boiling point tubes (10), boiling capillary (10), sample holder, cleaning tool, CD demo melting point monitor, protective cover, USB cable (2m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10499094 Supplier BUCHI Supplier No. 051999
Add to basket
Add to basket
Includes
100 melting capillaries, 10 boiling tubes, 10 boiling capillaries, calibration set M-560/565, sample holder, packing wire and cleaning tool.
Fast and accurate. BUCHI M-560/565 Melting Point Instruments have simple menus and programs that are easy to navigate on the intuitive LCD display, with either manual or automatic, real time video instrument.
  • Fail-safe calibration with automated determination and storage of calibration results
  • Automatic determination of melting and boiling points
  • Highly accurate temperature regulation and measurement
  • Optimized reproducibility with homogeneous packing of the capillary using the Sample Loader M-569
  • Control of almost any type of sample by measuring melting point, boiling point or slip melting point.
  • Double check for difficult samples by comparing manual and automated measurement
  • Observation and replay of the phase transition using color display and video recording
  • PC software included for easy management of methods and results
  • Compliant with Pharmacopeia methods (European, USP, Japanese)
  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.
Certifications
CE, UL, CSA
TRUSTED_SUSTAINABILITY
Specifications
Product Type Manual Melting Point Apparatus
Hertz 50/60 Hz
Voltage 100/240 V
Wattage 150 W
Accuracy ±0.3°C
Certifications/Compliance CE, UL, CSA
Depth (English) 14.5 in.
Depth (Metric) 37 cm
Display Type TFT
Electrical Requirements 100/240 V, 50/60 Hz
For Use With (Equipment) Determination of melting and boiling point
Height (English) 7.8 in.
Height (Metric) 20 cm
Includes Melting Point instrument
Width (English) 7.4 in.
Width (Metric) 19 cm