Learn More
3M™ Melamine Floor Pads, Gray/White
Click to view available options
Diameter (Metric):
330 mm
370 mm
406 mm
432 mm
505 mm
Unit Size:
Pack of 5
Description
- Floor pad for modern micro-porous floor surfaces such as ceramic, terracotta, marble, and concrete
- Cleans most dirt and marks with water alone, no cleaning additives or chemicals required
- Environmentally preferable
- Cleans up to 1000 m2 for single disc cleaning machines and 10,000 m2 for scrubber driers
Specifications
Specifications
| Product Type | Floor Pads |
| Color | Gray, White |
| Diameter (Metric) | 505 mm |
| For Use With (Application) | Cleaning |
| For Use With (Equipment) | Porous Floor Surfaces |