Learn More
Mefenamic Acid, TRC
Product Code. 17991955
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991955 50 mg 50mg
17971315 1 g 1g
17954075 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17991955 Supplier Toronto Research Chemicals Supplier No. TRCM20505050MG
 Restricted Item
Estimated Shipment: 19-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61-68-7
Molecular Formula C15 H15 N O2
Molecular Weight (g/mol) 241.29
Synonym Anthranilic acid, N-2,3-xylyl- (8CI), 2-[(2,3-Dimethylphenyl)amino]benzoic acid, 2-(2,3-Dimethylanilino)benzoic acid, 2',3'-Dimethyl-N-phenylanthranilic acid, Bafameritin M, Baphameritin M, Bonabol, C.I. 473, CN 35355, Coslan, INF 3355, In-M, Lysalgo, Mefacit, Mefenacid, Mefenamic acid, Mephenamic acid, N-(2,3-Dimethylphenyl)anthranilic acid, N-(2,3-Xylyl)-2-aminobenzoic acid, N-(2,3-Xylyl)anthranilic acid, NSC 94437, Namphen, Parkemed, Ponalar, Ponstan, Ponstel, Ponstil, Ponstyl, Pontal, Relafan, Tanston, Vialidon
IUPAC Name 2-(2,3-dimethylanilino)benzoic acid
SMILES Cc1cccc(Nc2ccccc2C(=O)O)c1C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)
Formula Weight 241.1103
Chemical Name or Material Mefenamic acid