Learn More
Corning™ Medium 199 (Mod.)
Description
- Complete line of media and reagents for mammalian in vitro cell culture
- Ready-to-use media is supplied in polyethylene teraphthalate (PET) plastic bottles
- Sonically welded cap liners eliminate toxic resins or glues that could leach into media
- Shipped at ambient temperature
Specifications
Specifications
| Concentration | 1 X |
| Product Type | Medium 199 |
| For Use With (Application) | To assess growth promotion and cytotoxicity characteristics |
| Quantity | 6/Pk. |
| Endotoxin Level | <0.25EU/ mL |
| Formulation | L-Glutamine, Earleâ€TMs Salts |
| pH | 7 to 7.4 |
| Content And Storage | 2° to 8°C |