Learn More
Pal International™ MediPal Alcohol-Free Disinfectant Wipes Canister
Product Code. 15104439
Product Code. 15104439 Supplier Pal International™ Supplier No. W615110MPCE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
2 Liter tub of 200 wipes. Pal International™ MediPal Alcohol-Free Disinfectant Wipes Canister is suitable for general everyday cleaning and disinfection of non-porous hard surfaces and non-invasive medical devices.
  • 2.0L Canister of 200 alcohol-free disinfecting wipes
  • Suitable for general everyday cleaning and disinfection of non-porous hard surfaces and non-invasive medical devices
  • Store on trolleys, countertops or wall-mounted in a dispenser bracket
  • Supports healthcare institutions to achieve both their clinical and budgetary targets
  • Help infection prevention teams to fight infection and ensure healthy, clinically clean environments for patients
  • Highly effective from 30 seconds
  • Proven to kill most microorganisms such as bacteria, viruses and spores
  • Independently tested to a range of European standards including the new European test EN16615:2015 (4-field test)
Product Identifier
  • Medipal Disinfectant Wipes
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains Polyhexamethylene Biguanide Hydrochloride. May produce an allergic reaction.
Precautionary Statement