Learn More
Pal International™ MediPal Alcohol-Free Disinfectant Wipes Canister
Description
- 2.0L Canister of 200 alcohol-free disinfecting wipes
- Suitable for general everyday cleaning and disinfection of non-porous hard surfaces and non-invasive medical devices
- Store on trolleys, countertops or wall-mounted in a dispenser bracket
- Supports healthcare institutions to achieve both their clinical and budgetary targets
- Help infection prevention teams to fight infection and ensure healthy, clinically clean environments for patients
- Highly effective from 30 seconds
- Proven to kill most microorganisms such as bacteria, viruses and spores
- Independently tested to a range of European standards including the new European test EN16615:2015 (4-field test)
Safety and Handling
Product Identifier
- Medipal Disinfectant Wipes
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains Polyhexamethylene Biguanide Hydrochloride. May produce an allergic reaction.
- P273-Avoid release to the environment.