Learn More
Fisherbrand™ Media Bottle PET With Cap
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 24
Pack of 48
Pack of 60
Pack of 96
Beskrivning
- Fisherbrand Media Bottles - These bottles are engineered to provide a comprehensive solution for storing media, biological buffers and a variety of reagents
- These bottles are designed to be leak-proof, shatter proof and freezer-safe, making them ideal for storing and transporting valuable samples
- Fisherbrand Media Bottles are the perfect choice for a reliable and convenient storage solution that meets the rigorous demands of modern laboratories
- Fisherbrand's media bottles are the perfect blend of durability, safety and versatility
Specifikationer
Specifikationer
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 0.264 gal. |
| Color | Clear |
| Material | PET |
| Sterility | Sterile |
| Closure Included | Yes |
| Disposable | Single-use |
| Graduated | Yes |
| Mouth | Threaded |
| Shape | Square |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering