Learn More
Meconin-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Meconin-d3 |
| CAS | 29809-15-2 |
| Molecular Formula | C10H7D3O4 |
| Quantity | 5 mg |
| Synonym | Phthalide, 6-methoxy-7-methoxy-d3- (8CI), 6-Methoxy-7-(trideuteriomethoxy)-3H-2-benzofuran-1-one, Meconin-d3, 6,7-Dimethoxy-1(3H)-isobenzofuranone-d3, Meconin-D3 (7-Methoxy-D3) |
| InChI Formula | InChI=1S/C10H10O4/c1-12-7-4-3-6-5-14-10(11)8(6)9(7)13-2/h3-4H,5H2,1-2H3/i2D3 |
| SMILES | [2H]C([2H])([2H])Oc1c(OC)ccc2COC(=O)c12 |
| Molecular Weight (g/mol) | 197.2 |
| Formula Weight | 197.0767 |