Learn More
Meconin-d3, TRC
Product Code. 30479148
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479148 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30479148 Supplier Toronto Research Chemicals Supplier No. TRCM2029022.5MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Meconin-d3
CAS 29809-15-2
Molecular Formula C10H7D3O4
Quantity 2.5 mg
Synonym Phthalide, 6-methoxy-7-methoxy-d3- (8CI), 6-Methoxy-7-(trideuteriomethoxy)-3H-2-benzofuran-1-one, Meconin-d3, 6,7-Dimethoxy-1(3H)-isobenzofuranone-d3, Meconin-D3 (7-Methoxy-D3)
InChI Formula InChI=1S/C10H10O4/c1-12-7-4-3-6-5-14-10(11)8(6)9(7)13-2/h3-4H,5H2,1-2H3/i2D3
SMILES [2H]C([2H])([2H])Oc1c(OC)ccc2COC(=O)c12
Molecular Weight (g/mol) 197.2
Formula Weight 197.0767