Learn More
SI Analytics™ Measuring Stand AVS™/HT
Product Code. 10365491 Shop All SI Analytics Products
Change view
Click to view available options
Temperature Range (Metric):
-80°C to +200°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10365491 -80°C to +200°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10365491 Supplier SI Analytics™ Supplier No. 285410954
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
SI Analytics™ Measuring Stand AVS™/HT is designed to be used with high temperatures of up to 200°C.
  • Constructed of anodised aluminum
  • Can withstand temperatures of up to 200°C
TRUSTED_SUSTAINABILITY
Specifications
Product Type Measuring Stand AVS/SHT
Temperature Range (Metric) -80°C to +200°C