Learn More
Hirschmann™ Class B Volumetric Flask with Stopper
Description
- Acc. to DIN EN ISO 1042
- PP
- Adjusted ringmark
- translucent
- with standard ground poly stoppers
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Polypropylene |
| Capacity (Metric) | 250 mL |
| Height (Metric) | 235 mm |
| No. per Pack | 5/Pack |
| Tolerance | ± 0.3 mL |
| Class | B |
| Bottom Shape | Round |
| Closure Type | Stopper |
| Closure Size | NS 19/26 |