Learn More
Lovibond™ MD 100 Chlorine Duo
Product Code. 12651705
Change view
Click to view available options
Temperature (Metric) Operating:
5°C to 40°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Operating unitSize
12651705 5°C to 40°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12651705 Supplier Lovibond™ Supplier No. 276020
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Lovibond MD 100 Chlorine Duo, Applications: Testing in the field, laboratory or plant room, Detection Range: 0 to 6mg/L, 0 to 10mg/L, 0 to 2mg/L, 0.1 to 8mg/L, Display Type: LCD, Filling Solution Type: Reagent
Specifications
Type MD 100 Chlorine Duo
Detection Range 0 to 6 mg/L, 0 to 10 mg/L, 0 to 2 mg/L, 0.1 to 8 mg/L
Display Type LCD
Filling Solution Type Reagent
Temperature (Metric) Operating 5°C to 40°C
For Use With (Application) Testing in the field, laboratory or plant room
Includes 4 Micro batteries (AAA), 3 round vials (Glass) with lids, tablets reagents and/or liquid reagents or VARIO powder reagent, guarantee sheet, certificate (COC), instruction manual
Warranty 5 or 10 Year
Length (Metric) 155 mm
Width (Metric) 75 mm
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
  • H372-Causes damage to organs.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.