Learn More
Zeiss™ MC 1000 Controller
Product Code. 11845123
Click to view available options
Type:
Controller MC 1000
Power Supply for VisiLED Illuminator
Unit Size:
Each
Product Code. 11845123
Brand: Zeiss™ 4170959040000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with VisiLED illuminator
Controller MC 1000 for VisiLED (D)
  • With 3 rotating knobs for dimming, segmentation (4 modes) and rotation
  • For control of one VisiLED illuminator
Power supply for VisiLED (D)
  • 100/240V
  • ,50/60Hz
Specifications
Type Controller MC 1000
For Use With (Equipment) VisiLED Illuminator
Quantity 1 Ea.