missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MBNL1 (Human) Recombinant Protein
Human MBNL1 full-length ORF ( AAH50535, 1 a.a. - 343 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004154-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGRCSRENCKYLHPPPHLKTQLEINGRNNLIQQKNMAMLAQQMQLANAMMPGAPLQPVPMFSVAPSLATNASAAAFNPYLGPVSPSLVPAEILPTAPMLVTGNPGVPVPAAAAAAAQKLMRTDRLEVCREYQRGNCNRGENDCRFAHPADSTMIDTNDNTVTVCMDYIKGRCSREKCKYFHPPAHLQAKIKAAQYQVNQAAAAQAAATAAAMTQSAVKSLKRPLEATFDLGIPQAVLPPLPKRPALEKTNGATAVFNTGIFQYQQALANMQLQQHTAFLPPGSILCMTPATSVVPMVHGATPATVSAATTSATSVPFAATATANQIPIISAEHLTSHKYVTQM
Specifications
AAH50535 | |
muscleblind-like (Drosophila) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBNL1 | |
GST |
4154 | |
Wheat germ expression system | |
10 μg | |
DKFZp686P06174, EXP, EXP35, EXP40, EXP42, KIAA0428, MBNL | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |