missing translation for 'onlineSavingsMsg'
Learn More

MBD6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA558606

Additional Details : Weight : 0.02000kg

Product Code. 15766481

  • 514.00€ / 100µL
Estimated Shipment: 20-05-2024
to see stock.
Add to basket

Description

Description

Immunogen sequence: MERSPRRTHH WQHNGELAEG GAEPKDPPPP GPHSEDLKVP PGVVRKSRRG RRRKYNPTRN SNSSRQDITL EPSPTARAA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Binds to heterochromatin. Does not interact with either methylated or unmethylated DNA (in vitro).
Specifications

Specifications

MBD6
Polyclonal
Unconjugated
MBD6
D10Wsu93e; KIAA1887; MBD6; methyl-CpG binding domain protein 6; methyl-CpG-binding domain protein 6; Methyl-CpG-binding protein MBD6
Rabbit
Antigen affinity chromatography
RUO
114785
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96DN6
MBD6
Recombinant protein corresponding to Human MBD6
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only