Learn More
Thermo Scientific™ MaxQ™ 4000 Gassing Manifold
Product Code. 12914115 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
MaxQ 4000
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12914115 MaxQ 4000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12914115 Supplier Thermo Scientific™ Supplier No. SHK40008GM
Only null left
Add to basket
Add to basket
Enable direct gassing into flasks for anaerobic cultures with the Thermo Scientific™ MaxQ™ 4000 Gassing Manifold.
Enable direct gassing into flasks for anaerobic cultures with the Thermo Scientific™ MaxQ™ 4000 Gassing Manifold. With exceptional innovation, reliability and accuracy, MaxQ™ 4000 shakers, platforms and accessories are the optimal choice for a busy lab.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gassing Manifold
Description Gassing manifold with 8 outlets
For Use With (Equipment) MaxQ 4000