Learn More
3M™ Maxim™ DX Safety Glasses
Description
- Safety spectacles with aspherical lens for 180° of Pure Vision and superior eye coverage
- DX coating provides enhanced anti-scratch and anti-fog protection
- Spectacles with ventilation channel across the lens to minimize misting
- Design provides excellent coverage and good field of vision
- Pliable, adjustable nose-pads and a 'soft-touch' brow guard
- Adjustable temple lengths and pantoscopic (angle) adjustments
Specifications
Specifications
| Certifications/Compliance | EN 166: EN 169 |
| Lens Type | DX |
| Lens Tint | Shade 5 |
| Product Type | Safety glasses |
| For Use With (Application) | Construction |
| Frame Color | Black, Gray |
| Lens Material | Polycarbonate |