Learn More
3M™ Maxim™ Ballistic Lens Utility Pack
Click to view available options
:
Frame Color: Black
Description
- Safety spectacles with 3 lens color options for different light and target conditions
- Frame nose pads and pantoscopic angle are fully adjustable and pliable soft-touch materials are used in key contact points
- Design provides excellent coverage and good field of vision
- Meets the stringent STANAG 2920 ballistic test
- Temples feature adjustable pantoscopic angle
- Lens coatings provide excellent scratch resistance and anti fogging
Specifications
Specifications
| Lens Type | DX |
| Lens Tint | Clear, Bronze, Amber |
| Product Type | Ballistic Lens Utility Pack |
| Frame Color | Black |
| Includes | Three lenses |