missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas Maxi Plus Cooled Gel Electrophoresis Unit
Description
- Cooled base to allow separations to be performed faster, higher voltage,without loss of resolution
- Cooling ports to connect to laboratory water supply or optional external chiller
- Twelve comb slots
- Multi-channel pipette compatible combs
- Color coded, height adjustable combs
- Buffer recirculation ports to connect to a peristaltic pump
- Coloured loading strips
- UV-transparent acrylic casting tray
- Side handles for safe and easy transportation around laboratory
Comb Specifications:
- Power output connector: Shrouded, 4mm (diameter)
- Comb thickness: 1, 1.5 or 2mm
- Comb throughput: 26 to 52 samples
- Comb slots: 12
- Migration distance between comb slots: 2.0cm
Specifications
Specifications
| Type | Gel Electrophoresis Unit |
| Gel Dimensions | 300 x 250 mm |
| Includes | 1 x gel casting tray, 6 x 1.0mm 26 sample combs, buffer recirculation ports, power supply connectors and loading strips |
| Model | Maxi-Plus |
| Sample Size | 624 (max.) |
| Volume (Metric) Buffer | 3000 mL |
| Dimensions (L x W) | 30 x 25 cm (Gel) |
| Dimensions (L x W x H) | 56 x 33 x 9 cm |
| Length (Metric) Gel | 300 mm |
| Width (Metric) Gel | 250 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?