Learn More
Brady™ Matte Polyester RFID Chemical Resistant Slide Labels
Product Code. 17445993
Change view
Click to view available options
Color:
White
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17445993 White 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17445993 Supplier Brady™ Supplier No. 309083
Only null left
Add to basket
Add to basket
UHF RFID labels for use on medical slides.
  • For use in pharmaceutical industry and healthcare
  • Designed for use on medical slides in the laboratory environment
  • Polyester tag face material with chip bonded to aluminum antenna using anisotropic conductive film adhesive
  • Suitable for thermal transfer printing, including RFID encoding
  • Labels are outside wound and 100% inspected, 500 good RFID labels per roll with bad tags marked
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) RFID Labeling, Healthcare and Hospital Labeling, Laboratory Labeling
For Use With (Equipment) Sato CL4NX Plus Industrial Label Printer, Zebra™ ZT411 Industrial Label Printer
Labels per Row 1
Min. Operating Temperature (Metric) -80°C
No. per Pack 500 Labels/Roll
Quantity 500 Labels
Temperature Range (English) -112°F to 212°F
Temperature Range (Metric) -80°C to +100°C
Thickness (English) 0.0114 in.
Thickness (Metric) 0.29 mm
Vertical Repeat (Metric) 20 mm
Dimensions (L x W) 16 x 24 mm
Length (English) 0.63 in.
Width (English) 0.94 in.
Width (Metric) 24 mm
Height (English) 0.63 in.
Height (Metric) 16 mm
Alternate Part Number THT-UHF-481-24x16