Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
GREENER_CHOICE
Product Code. 11902261
Change view
Click to view available options
Color:
Black
Blue
Colorless
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Screw Cap
Screw Cap Insert
Screw Cap Tray
Screw Cap Tray, Empty
Unit Size:
Pack of 480
Pack of 5
Pack of 500
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
11902261 Black Screw Cap Pack of 500
11932261 Orange Screw Cap Pack of 500
11942261 Pink Screw Cap Pack of 500
11865482 Gray Screw Cap Tray Pack of 480
11875482 Pink Screw Cap Tray Pack of 480
10736554 Colorless Screw Cap Pack of 500
11632000 Red Screw Cap Pack of 500
10780140 Yellow Screw Cap Pack of 500
11681970 Green Screw Cap Pack of 500
10401908 White Screw Cap Pack of 500
11651990 Purple Screw Cap Pack of 500
10040273 Red Screw Cap Insert Pack of 500
11577429 Yellow Screw Cap Tray, Empty Pack of 5
12865601 Colorless Screw Cap Tray Pack of 480
11642000 Red Screw Cap Tray Pack of 480
11612010 Yellow Screw Cap Tray Pack of 480
10290740 Blue Screw Cap Tray Pack of 480
11317201 Green Screw Cap Tray Pack of 480
11895482 White Screw Cap Tray Pack of 480
11377201 Purple Screw Cap Tray Pack of 480
11601970 Blue Screw Cap Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 11902261 Supplier Thermo Scientific™ Supplier No. 4470BLK
  • ONLINE EXCLUSIVE PRICE
    187.00€
    182.99€ / Pack of 500
    Save 4.01€
    2% Off
View all promotions
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Superior Cap Design
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -196ºC
Cap Options Simplify Sample Identification
  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors
All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
TRUSTED_SUSTAINABILITY
Specifications
Product Type Screw Cap
Material Polypropylene
Color Black
Packaging Bulk, 500 Caps
Sterility Sterile
For Use With (Equipment) ScrewTop Tube
Fit for Purpose Sterile and PCR Ready