Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
Click to view available options
Capacity (Metric):
25 mL
75 mL
100 mL
Material:
Polypropylene
Polystyrene
Packaging:
10 Bags of 10
10 pcs /bag
50 Individually wrapped
Type:
Reagent Reservoir
Reagent Reservoir with Divider
Reservoir Lid
Dimensione della confezione:
Pack of 10
Pack of 100
Pack of 50
Descrizione
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
- Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Specifica
Specifica
| Color | White |
| Capacity (Metric) | 75 mL |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reservoir Lid |
| No. per Case | 10 |
| No. per Pack | 10 |
| Packaging | 10 pcs /bag |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto