Learn More
IKA™ MATRIX Attachment
Product Code. 16208691 Shop All IKA Products
Click to view available options
Holds:
Conical Vials
Reaction Vessels
Tube Capacity:
0.5 mL
1.5 mL
2.0 mL
5.0 mL
15 mL
50 mL
No. of Positions:
4
8
24
Unit Size:
Each
Product Code. 16208691
Brand: IKA™ 0030000834
Only null left
Add to basket
Add to basket
Attachment design ensures an optimal temperature transfer to guarantee valid results.
  • Thermoshakers automatically recognize which attachment is connected to the device via an interface in the locking mechanism
  • Attachments can be changed quickly and easily with an ergonomic lever
  • As part of IKA's Integrated Safety Concept, the insulating plastic coating of the attachment ensures user safety, as high temperatures are not transferred to the frame
  • Suitable for use with MATRIX Orbital Delta Plus, and MATRIX Delta Plus
TRUSTED_SUSTAINABILITY
Specifications
Holds Reaction Vessels
Tube Capacity 2.0 mL
Product Type Attachment for Reaction Vessels
Dimensions (L x W x H) 150 x 141 x 53 mm
For Use With (Application) Blood Samples, Pharmaceutical Agents,/DNA/RNA Samples
For Use With (Equipment) MATRIX Orbital Delta Plus, MATRIX Delta Plus
No. of Positions 24