Learn More
Thermo Scientific™ Matrix™ ScrewTop Tri-coded Tubes
BIOPHARMA
Product Code. p-9374001
Change view
Click to view available options
Capacity (Metric):
0.64 mL
1.0 mL
1.1 mL
Sterility:
Non-sterile
Sterile
Product Type:
Screw Top Tube
Screw Top with White Patch
Unit Size:
Pack of 480
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Product Type Unit Size
15389804 0.64 mL Sterile Screw Top Tube Pack of 480
15480004 0.64 mL Sterile Screw Top with White Patch Pack of 480
13415868 0.64 mL Sterile Screw Top with White Patch Pack of 480
15573724 0.64 mL Sterile Screw Top Tube Pack of 480
13466898 0.64 mL Sterile Screw Top with White Patch Pack of 480
15389784 1.1 mL Sterile Screw Top Tube Pack of 480
15419973 1.1 mL Sterile Screw Top with White Patch Pack of 480
12874953 1.0 mL Sterile Screw Top with White Patch Pack of 480
13466888 1.1 mL Sterile Screw Top with White Patch Pack of 480
15593704 1.1 mL Sterile Screw Top Tube Pack of 480
17258820 0.64 mL Non-sterile Screw Top with White Patch Pack of 480
17268820 0.64 mL Non-sterile Screw Top with White Patch Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15389804 Supplier Thermo Scientific™ Supplier No. 3743WP1D
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μl and 1.0mL ScrewTop Tubes.
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 0.64 mL  and 1.1 mL ScrewTop Tubes. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D,1D linear and human readable barcode. These tubes can be ordered to come in racks (barcoded or non-barcoded) or may be purchased in bulk. The tri-coded tubes can be purchased sterile or non-sterile. We also offer tubes with white patch only if your work does not require the 1D linear and human readable code. The 2D screwtop tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment.
Dependable Sample Protection
  • Tubes are constructed of virgin class VI medical grade polypropylene
  • ScrewTop caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy ABS to protect tubes in case of drops
  • ScrewTop tubes are compatible with storage down to -196°C
Secure Tracking
  • Permanent, unique 2D tube barcode is laser-etched on bottom of each tube
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking
  • 2D barcode on tubes allow you to scan and decode tubes all at once without removing them from the rack, and allows data to be associated with each tube
  • Tube and rack are compatible with Thermo Scientific™ VisionMate™ readers
Superior Storage Format
  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping / decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at once
Images
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Description ScrewTop Tubes in Barcoded Latch Racks
Closure Color Colorless
Autoclavable Not Autoclavable
Capacity (Metric) 0.64 mL
Bottom Shape Conical
Material Polypropylene
Packaging Bulk, 480 Tubes
Barcode Linear barcode
Product Line Matrix
Sterility Sterile
Capacity (English) 0.0216 fl oz.
Fit for Purpose Sterile and PCR Ready
Product Type Screw Top Tube
Green Features ACT Ecolabel (Gold)