Learn More
Tork Matic™ Paper Hand Towel Roll H1
GREENER_CHOICE
Product Code. p-200113548 Shop All Tork Products
Click to view available options
Color:
Blue
Natural
Unit Size:
6 rolls
Product Code. 30245604
Brand: Tork 290099
Only null left
Add to basket
Add to basket
The rolls are suitable for the Tork Matic™ Hand Towel Roll Dispenser, developed for easy maintenance in high traffic washrooms.
  • Suit the needs of frequently visited washrooms with this Advanced quality paper towel roll.
  • Reduce waste and boost hygiene with one-at-a-time dispensing from a compatible Tork paper towel dispenser.
  • Balance cost-effectiveness with great hand-drying performance with these advanced quality hand towels.
  • Commit to better environmental practices with this EU Ecolabel certified product.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Towel Roll
Length (English) 164.04 yd.
Length (Metric) 150 m
Width (English) 8.27 in.
Width (Metric) 21 cm
Material Paper
Color Natural
Packaging Quantity 6/Cs.
Ply 2-ply