Learn More
Masterflex™ L/S™ Easy-Load™ Pump Head Mounting Hardware
Easy stack multiple pump heads on your Masterflex™ L/S™ drive
63.25€ - 89.30€
Specifications
For Use With | Masterflex L/S Easy Load Pump Heads |
---|---|
Material | Stainless Steel |
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
11857903
|
Masterflex™
07013-04 |
SS mounting hardware; mounts 1 L/S Easy-Load pump head | 4/Pk. |
63.25€
Pack of 4 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11775079
|
Masterflex™
07013-05 |
SS mounting hardware; mounts 2 L/S Easy-Load pump heads | 4/Pk. |
67.80€
Pack of 4 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
11785079
|
Masterflex™
07013-08 |
SS mounting hardware; mounts 3 L/S Easy-Load pump heads | 1/Ea. |
80.20€
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11755089
|
Masterflex™
07013-09 |
SS mounting hardware; mounts 4 L/S Easy-Load pump heads | 1/Ea. |
89.30€
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
Description
Easy stack multiple pump heads on your Masterflex™ L/S™ drive
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
- Suitable for use with one Easy-Load™ pump head
Specifications
Masterflex L/S Easy Load Pump Heads | |
Stainless Steel |