Learn More
Masterflex™ L/S™ Power Supply, Dual Voltage 115/230 VAC to 12 VDC, 12-ft Cable
Product Code. 11826723 Shop All Masterflex Products
Change view
Click to view available options
Electrical Requirements:
115/230 to 12 VDC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Unit Size
11826723 115/230 to 12 VDC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11826723 Supplier Masterflex™ Supplier No. 7720007
  • ONLINE EXCLUSIVE PRICE
    261.00€
    251.09€ / Pack of 1
    Save 9.91€
    3% Off
Estimated Shipment: 07-12-2026
Make the most of your Masterflex™ drives replacement parts and other accessories
Suitable for use with 07571-00 L/S™ drive
Specifications
Product Type Power Supply
Voltage 115/230 to 12 V
Electrical Requirements 115/230 to 12 VDC