Learn More
Masterflex™ Platinum-Cured Silicone Tubing
Product Code. 10128791
Click to view available options
Tubing Size:
73
Unit Size:
7.5m
Product Code. 10128791
Brand: Masterflex™ MF0316
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure optimal performance from your Masterflex™ pump. Masterflex™ Platinum-Cured Silicone Tubing is manufactured to extremely close tolerances and ideal for pharmaceutical and biotechnological use.
  • Slightly greater clarity than peroxide-cured tubing.
  • Smooth surfaces; lower protein binding levels.
  • Fewer potential leachables.
  • Ideal for pharmaceutical and biotechnology use.
Specifications
Diameter (Metric) Inner 9.5 mm
Material Silicone (Platinum-Cured)
Min. Operating Temperature (Metric) -50°C
Tubing Size 73
Flow Rate 8000 mL/min maximum
Width (Metric) 7.6 m
Maximum Pressure 40psi, 2.7 bar
Vacuum Level 660 mm Hg maximum
Max. Operating Temperature (Metric) 230°C