Learn More
Masterflex™ DB25 Male Connector
Product Code. 11755369
Click to view available options
For Use With:
20 to 22 gauge Shielded Wire
Unit Size:
Each
Product Code. 11755369
Brand: Masterflex™ 0752394
Create your own cable for optimization of your Masterflex™ drive
Choose the correct connection for your Masterflex™ pump
Specifications
For Use With (Application) For creating your own cable
For Use With 20 to 22 gauge Shielded Wire
Quantity 1 Each
Type DB25 Male Connector