Learn More
Masterflex™ Chem-Durance™ Bio L/S™ High-Performance Precision Pump Tubing
Product Code. 11836953
Click to view available options
Tubing Size:
15
24
35
36
Unit Size:
Each
Product Code. 11836953
Brand: Masterflex™ 0644235
Use this tubing for a wide range of laboratory, pharmaceutical processing and biotech applications. Masterflex™ Chem-Durance™ Bio L/S™ High Performance Precision Pump Tubing features excellent chemical resistance and durability under pressure.
  • For applications involving pressure, suction lift, viscous fluids or long tubing life
  • Thicker wall compared to precision pump tubing
  • Performs in a wide range of chemical applications limiting the number of tubing formulations you need to keep on hand
  • Low spallation maintains purity of fluid
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers and extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids Complies with USP Class VI, FDA, REACH, RoHS
  • Excellent chemical compatibility and biocompatibility
  • Excellent pump life and durability under pressure
  • Excellent chemical resistance and long pumping life
  • Meets USP Class VI requirements
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 7.9 mm
Material Chem-Durance Bio
Overall Length (Metric) 1520 cm
For Use With (Application) Laboratory, pharmaceutical processing and biotech applications
Min. Operating Temperature (Metric) -60°C
Tubing Size 35
Flow Rate 3.8 to 2300 mL/min.
Height (Metric) 8.8 m
Color Opaque
Maximum Pressure 35 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type Pump Tubing