Learn More
Masterflex™ Tygon™ Fuel & Lubricant I/P™ Precision Tubing
Product Code. 11795099
Click to view available options
Tubing Size:
73
82
Unit Size:
Each
Product Code. 11795099
Brand: Masterflex™ 0640173
Only null left
Add to basket
Add to basket
Use this tubing for applications involving fuels and industrial lubricants. Masterfelx™ Tygon™ Fuel & Lubricant I/P™ Precision Tubing is deal for transferring gasoline, kerosene, heating oils, cutting compounds and glycol-based coolants.
  • Resists most hydrocarbons, embrittlement and swelling
  • Very low gas permeability
  • Complies with USP Class V, EP, FDA, REACH, RoHS, ADCF
  • Ozone- and UV-resistant
  • Low extractability
  • Translucent yellow High dielectric constant
  • Thermoplastic 
  • PVC-based material with plasticizer
Specifications
Diameter (Metric) Inner 9.5 mm
Material Thermoplastic
Overall Length (Metric) 1500 cm
For Use With (Application) transferring Hydrocarbons, Gasoline, Kerosene, heating oils, cutting compounds, and Glycol based coolants
Min. Operating Temperature (Metric) -37°C
Packaging Quantity 1 Each
Tubing Size 73
Flow Rate 0.01 to 8L/min.
Hose Barb 3/8in.
Height (Metric) 8.8 m
Gas Permeability Low
Color Translucent Yellow
Maximum Pressure 40 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type Pump Tubing