Learn More
Atago™ MASTER-BC Model Battery Coolant Checker Refractometer
Description
- For use with chemicals found in antifreeze mixtures in automobiles, thermal catalysts for solar power systems and other industrial applications
- MASTER-BC is the successor model to the conventional BC-2E model
- Scale is graduated in °C
- Constructed of durable material suitable for salty and acid samples
Specifications
Specifications
| Type | Battery Coolant Refractometer |
| Scale Range | 1.150 to 1.300 Battery Fluid d20/20 |
| Model | MASTER-BC |
| Scale Type | Freezing temperature, Battery fluid d20/20 |
| For Use With (Application) | Measuring specific gravity of battery fluid and checking the freezing temperature of ethylene glycol and propylene glycol which are used as antifreeze mixture in automobiles |
| Weight (Metric) | 90 g |
| Temperature Range (Metric) | 0 to -50°C Ethylene Glycol/Propylene Glycol |