Learn More
Atago™ MASTER-93H Model Refractometer
Product Code. 10795973
Change view
Click to view available options
Scale Range:
45 to 93 Brix
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scale Range Unit Size
10795973 45 to 93 Brix Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10795973 Supplier Atago™ Supplier No. 2374
  • ONLINE EXCLUSIVE PRICE
    193.00€
    174.71€ / Pack of 1
    Save 18.29€
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Use this heat resistant refractometer for measuring high-temperature samples at their respective production sites. Atago™ MASTER-93H Model Refractometers feature a prism made of corrosion resisting tempered glass for longer service life.
  • Best suited for measuring jam, marmalade and candy
  • Easy to read and  hold
  • Designed for smooth sampling
  • Easy disposal of sample from the edge
  • Balanced structure
  • Streamlined body and the strong grip with rich texture
Specifications
Type Refractometer
Scale Range 45 to 93 Brix
Model MASTER-93H
Accuracy ±0.5% Brix at 10 to 40°C
Repeatability ±0.25% Brix
Scale Type Brix
For Use With (Application) Measuring jam, marmalade candy
Weight (Metric) 130 g