Learn More
Watson-Marlow Marprene Double Segment Manifold Tubing
Click to view available options
Diameter (Metric) Inner:
1.29 mm
Unit Size:
Pack of 6
Description
- Marprene is a long life thermoplastic elastomer with resistance to a wide range of chemicals.
- It lasts at least 10 times as long as tubing in other materials.
- Long-life tubing means stoppages for a tube change are rare events, easily fitted in with general maintenance.
- Marprene has wide chemical compatibility, and is highly resistant to oxidizing agents such as ozone, peroxides and sodium hypochlorite.
- Marprene is beige and opaque to visible and ultraviolet light, with low permeability to oxygen, carbon dioxide and nitrogen.
- It meets USDA standards for food handling.
Specifications
Specifications
| Diameter (English) Inner | 0.05 in. |
| Diameter (Metric) Inner | 1.29 mm |
| Material | Marprene |
| Overall Length (English) | 1.5 ft. |
| Overall Length (Metric) | 0.46 m |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.8 mm |
| Color | Grey/Grey |
| Product Type | Double Segment Tubing |