Description
- Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many other chemicals
- Integrated gripping edge on funnel lid increases grip without having to touch possibly contoured lid
- Innovative funnel lid with integrated sealing ring closes funnel edge to prevent escape of explosive or health-endangering gases
- Sturdy hinge guarantees long service life of the closing mechanism
- Rotating screw cap makes it easier to screw lid funnel on and off the canister
- Removable sieve for easy cleaning
- Conductive version of MARCO V2.0 is black, differring in color from the non-conductive version
Specifications
Specifications
| Diameter (Metric)Top | 140 mm |
| Capacity (Metric) | 500 mL |
| Material | HDPE-EC |
| Height (Metric) | 125 cm |
| Color | Black |
| Includes | Removable Sieve, Earthing Cable |
| Stopper Number | S70/71 |
| Thread Size (Metric) | S70/71 |
| Product Type | Safety Funnel with Hinged Lid |