missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MAPK Phospho-Specific Array

Product Code. 16002445
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16002445 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16002445 Supplier Abnova™ Supplier No. AA0033.1Kit(2Samples)
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MAPK Phospho-Specific Array includes 185 highly specific and well-characterized phosphorylation antibodies in the MAPK pathway.

  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum

Specifications

Content And Storage 4°C
Detection Method Fluorometric
Quantity 1 Kit
For Use With (Application) Profiling
Product Type Array/Library
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.