Learn More
- Home
- Products
- Vials
- Vial Capping and Decapping Equipment
- Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 10231585| Product Code. | Size | Type | unitSize |
|---|---|---|---|
| 10231585 | 13 mm | Manual Crimper (Flip-off) | Each |
| 15280389 | 8 mm | Manual Crimper | Each |
| 14860742 | 11 mm | Manual Crimper | Each |
| 13488988 | 20 mm | Manual Crimper | Each |
| 10147664 | 8 mm | Manual Crimper | Each |
| 10429454 | 11 mm | Manual Crimper | Each |
| 10566765 | 13 mm | Manual Crimper | Each |
| 12400268 | 20 mm | Manual Crimper | Each |
| 10372525 | 20 mm | Manual Crimper (Flip-off) | Each |
| 10250535 | 28 mm | Manual Crimper | Each |
| 10773667 | 30 mm | Manual Crimper | Each |
| 10229964 | 32 mm | Manual Crimper | Each |
| 11528470 | 11 mm | Manual Crimper | Each |
| 11538470 | 20 mm | Manual Crimper | Each |
| 15290389 | 8 mm | - | Each |
| 15188784 | 20 mm | - | Each |
| 10259614 | 8 mm | Manual Decrimper | Each |
| 10488084 | 11 mm | Manual Decrimper | Each |
| 10240535 | 13 mm | Manual Decrimper | Each |
| 10279364 | 20 mm | Manual Decrimper | Each |
| 12852344 | 28 mm | Manual Decrimper | Each |
| 10301535 | 30 mm | Manual Decrimper | Each |
| 11528240 | 32 mm | Manual Decrimper | Each |
| 11564815 | 20 mm | Manual Crimper | Each |
| 10530303 | 11 mm | Manual Decrimper | Each |
| 10143362 | 20 mm | Manual Decrimper | Each |
| 12054907 | 11 mm | Manual Decapping Plier | Each |
| 10200205 | 30 mm | Manual Decapping Plier | Each |
Use promo code 28509
Log in to see stock.
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
- Factory set to propery crimp standard-thickness seals
- Adjustable handle setting scree allows proper crimps with septa of varying thickness
Specifications
Specifications
| Description | Manual Crimper, Flip-Off |
| No. per Pack | 1 Ea. |
| Type | Manual Crimper (Flip-off) |
| For Use With (Equipment) | National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial |
| Size | 13 mm |
| For Use With (Application) | Industrial Chromatography |
| Product Line | National |