Learn More
Watson-Marlow Manifold Tubing - Silicone
Product Code. 17867963
Click to view available options
Tubing Size:
0.63mm
0.76mm
0.88mm
1.02mm
1.14mm
1.42mm
1.52mm
1.65mm
1.85mm
2.05mm
2.79mm
Diameter (Metric) Inner:
0.63 mm
0.76 mm
0.88 mm
1.02 mm
1.14 mm
1.42 mm
1.52 mm
1.65 mm
1.85 mm
2.05 mm
2.79 mm
Color:
Black/Black
Blue/Blue
Green/Green
Orange/Orange
Orange/White
Purple/Purple
Purple/White
Red/Red
White
Yellow
Yellow/Blue
Unit Size:
Each
Pack of 6
Product Code. 17867963
Brand: Watson-Marlow 983.0063.000
Only null left
Add to basket
Add to basket
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 1/50 in.
Diameter (Metric) Inner 0.63 mm
Material Silicone
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 mm
For Use With 400DM2 and 400DM3 pump heads and multichannel cartridge
Tubing Size 0.63mm
Flow Rate 0.04 to 3.1 (four roller pump), 0.04 to 2.6 (eight roller pump)mL/min.
Color Orange/White
Product Type Pump Tubing