Learn More
Manganese(Iv) Oxide, activated, 85%, Honeywell™ Fluka™
Product Code. p-7120549 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
Product Code. 15642070
Brand: Honeywell-Fluka 2176465G
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Activated, ∼85%, <10 μm
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
Specifications
Quantity 5 g
UN Number NONH for all modes of transport
Formula Weight 86.94g/mol
Percent Purity 85%
Packaging Poly Bottle
Chemical Name or Material Manganese(Iv) Oxide