Learn More
Manganese(IV) Oxide, ≥90%, Reagent grade, Honeywell™
Product Code. p-7120548 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. 15633210
Brand: Honeywell Research Chemicals 310700500G
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
10 μm, Reagent Grade,≥90%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
Specifications
Melting Point 535°C
Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 86.94g/mol
Percent Purity ≥90%
Grade Reagent
Packaging Poly Bottle
Density 5.21g/cm3
Chemical Name or Material Manganese(Iv) Oxide