missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mandi

Product Code. 18777726
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777726 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18777726 Supplier Tocris Bioscience™ Supplier No. 7681/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly efficient chemical inducer of proximity (CIP)

Mandi is a chemical inducer of proximity (CIP) in vivo and in vitro (EC50 = 0.43 μM in a luciferase gene expression assay). Mandi induces complex formation between PYR1Mandi, a mutant of Abscisic Acid (Cat. No. 6554) receptor PYR1, and Abscisic Acid insensitive (ABI). Mandi may be used to induce protein colocalization and interaction. It is non-toxic, orthogonal to mammalian processes and other CIPs, and highly cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-N-[2-[3-methoxy-4-(2-propyn-1-yloxy)phenyl]ethyl]-α-(2-propyn-1-yloxy)benzeneacetamide
CAS 374726-62-2
Quantity 10 mg
Target Protein Dimerizers
Molecular Formula C23H22ClNO4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.