missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MammoCult™ Human Medium Kit
Description
- MammoCult™ Medium (Human) is a serum-free culture medium optimized for the culture of mammospheres from normal human primary breast tissues and tumorspheres from human breast cancer cell lines
- For preparation of complete MammoCult™ Medium, Hydrocortisone Stock Solution (Catalog #07925) and Heparin Solution (Catalog #07980) are also required
Specifications
Specifications
| Cell Type | Cancer Cells and Cell Lines, Mammary Cells |
| For Use With (Application) | Cell Culture, Maintenance, Spheroid Culture |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?