Learn More
Malt Extract, MP Biomedicals
Product Code. 11437582 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437582 500 g 500g
11427582 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11437582 Supplier MP Biomedicals Supplier No. 0215531590
From starch digestion. A source of carbohydrates
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8002-48-0
Specifications
Quantity 500 g
Chemical Name or Material Malt Extract