Learn More
Malonic Acid Disodium Salt Monohydrate, TRC
Product Code. 30480856
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480856 10 g 10g
30471420 5 g 5g
30482209 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480856 Supplier Toronto Research Chemicals Supplier No. TRCM15803010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26522-85-0
Molecular Formula C3H2Na2O4 . H2O
Molecular Weight (g/mol) 166.04
IUPAC Name disodium;propanedioate;hydrate
SMILES O.[Na+].[Na+].[O-]C(=O)CC(=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C3H4O4.2Na.H2O/c4-2(5)1-3(6)7;;;/h1H2,(H,4,5)(H,6,7);;;1H2/q;2*+1;/p-2
Formula Weight 165.9854
Chemical Name or Material Malonic Acid Disodium Salt Monohydrate