Learn More
Malonic Acid-1,2,3-13C3, TRC
Product Code. 30481824
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481824 100 mg 100mg
30483827 10 mg 10mg
30470208 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481824 Supplier Toronto Research Chemicals Supplier No. TRCM158009100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102342-85-8
Molecular Formula 13C3H4O4
Molecular Weight (g/mol) 107.04
Synonym Propanedioic Acid-1,2,3-13C3, 1,3-Propanedioic Acid-1,2,3-13C3, Carboxyacetic Acid-1,2,3-13C3, Dicarboxymethane-1,2,3-13C3, Methanedicarboxylic Acid-1,2,3-13C3, NSC 8124-2-1,2,3-13C3, [1,2,3-13C3]-Malonic acid
SMILES O[13C]([13C]([H])([H])[13C](O)=O)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)/i1+1,2+1,3+1
Chemical Name or Material Malonic Acid-1,2,3-13C3