Learn More
Malonaldehyde-1,3-d2 Bis(diethyl Acetal), CDN
Specifications
Specifications
| Chemical Name or Material | 1,1,3,3-Tetraethoxypropane-1,3-d2 |
| CAS | 105479-86-5 |
| Molecular Formula | C11 2H2 H22 O4 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C11H24O4/c1-5-12-10(13-6-2)9-11(14-7-3)15-8-4/h10-11 H,5-9H2,1-4H3/i10D,11 D |
| SMILES | [2 H]C(CC([2 H])(OCC)OCC)(OCC)OCC |
| IUPAC Name | 1,3-dideuterio-1,1,3,3-tetraethoxypropane |
| Molecular Weight (g/mol) | 222.32 |
| Formula Weight | 222.18 g/mol |
| Percent Purity | 96 atom % D, min 98% Chemical Purity |