Learn More
Cadence Science™ Malleable Stainless Steel Animal Feeding Needles
Click to view available options
Gauge:
18G
20G
Diameter (Metric) Ball:
1.9 mm
2.3 mm
Length (English):
1.5 in.
2 in.
3.1 in.
Unit Size:
Each
Pack of 100
Description
- Type 304 stainless steel construction
- Malleable
- Sterile
- Disposable
- 100/box, 20 x 5/sterile pouch
Specifications
Specifications
| Product Type | Animal Feeding Needle |
| Material | Stainless Steel (304) |
| Disposable | Single-use |
| For Use With (Application) | Animal feeding |
| Gauge | 18G |
| Sterility | Sterile |
| Length (English) | 3.1 in. |
| Diameter (Metric) Ball | 2.3 mm |